Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024919.1 | NP_001020090.1 | 4 | 1152 | ||
GenBank mRNA | AK009485.1 | - | 4 | 1134 | ||
BC116435.1 | - | 5 | 984 | 40104481 | ||
BC116436.1 | - | 5 | 984 | 40104482 | ||
BC128508.1 | - | 5 | 1054 | |||
BC128509.1 | - | 5 | 984 | 40104485 |