Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006872.4 | NP_006863.2 | 1, 2 | 118 | ||
GenBank mRNA | AF106857.1 | - | 1, 2 | 38 | ||
AK223350.1 | - | 1, 2 | 42 | |||
BC025991.1 | - | 1, 2 | 67 | 4826221 | ||
BC064585.1 | - | 1, 2 | 22 | |||
BG717289.1 | - | 1, 2 | 94 | |||
HY049561.1 | - | 1, 2 | 121 |