Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010153.1 | NP_034283.1 | 24 | 2855 | ||
GenBank mRNA | AY686636.1 | - | 24 | 2855 | ||
BC029028.1 | - | 370 | ||||
BC036180.1 | - | 459 | ||||
BC049279.1 | - | 667 | ||||
BC106091.1 | - | 3006 | 30844039 |