Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 41 | 4541 | ||
NM_014689.2 | NP_055504.2 | 41 | 4684 | |||
XM_005246673.4 | XP_005246730.1 | 41 | 4543 | |||
XM_006712617.3 | XP_006712680.1 | 41 | 4708 | |||
XM_017004441.1 | XP_016859930.1 | 41 | 4711 | |||
XM_017004442.1 | XP_016859931.1 | 41 | 4687 | |||
XM_017004443.1 | XP_016859932.1 | 41 | 4760 | |||
XM_017004444.1 | XP_016859933.1 | 40 | 4667 | |||
XM_017004445.1 | XP_016859934.1 | 41 | 4541 | |||
XM_017004446.1 | XP_016859935.1 | 40 | 4565 | |||
XM_017004447.1 | XP_016859936.1 | 41 | 4683 | |||
GenBank mRNA | AF250396.1 | - | 1 | 53 | ||
AK293847.1 | - | 7 | 610 | |||
AK293879.1 | - | 12 | 1090 | |||
AK307914.1 | - | 1 | 46 | |||
CR749492.1 | - | 16 | 1529 | |||
EU236710.2 | - | 41 | 4538 |