Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100621.2 | NP_001094091.1 | 3 | 538 | ||
NM_001100622.2 | NP_001094092.1 | 3 | 538 | |||
NM_001288770.1 | NP_001275699.1 | 3 | 699 | |||
NM_001288771.1 | NP_001275700.1 | 3 | 699 | |||
NM_017941.5 | NP_060411.2 | 3 | 538 | |||
XM_005257487.3 | XP_005257544.1 | 3 | 531 | |||
XM_006721966.3 | XP_006722029.1 | 3 | 538 | |||
XM_011524961.1 | XP_011523263.1 | 3 | 538 | |||
XM_011524962.2 | XP_011523264.1 | 2 | 436 | |||
XM_017024806.1 | XP_016880295.1 | 2 | 464 | |||
GenBank mRNA | AK000728.1 | - | 3 | 498 | ||
AK292838.1 | - | 3 | 467 | |||
AK301905.1 | - | 3 | 699 | |||
AL136740.1 | - | 3 | 505 | |||
AY163812.1 | - | 3 | 523 | |||
BC005005.2 | - | 2 | 443 | 3544728 |