Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RET | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020630.4 | NP_065681.1 | 2904 | |||
NM_020975.4 | NP_066124.1 | 2904 | ||||
GenBank mRNA | AK291807.1 | - | 2774 | |||
AK294827.1 | - | 2126 | ||||
BQ070075.1 | - | 425 |