Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017943.3 | NP_060413.2 | 2 | 1691 | ||
NM_152231.1 | NP_689417.1 | 2 | 1584 | |||
XM_005267791.4 | XP_005267848.1 | 2 | 1880 | |||
XM_006720185.3 | XP_006720248.1 | 2 | 1544 | |||
XM_017021391.1 | XP_016876880.1 | 2 | 1608 | |||
XM_017021392.1 | XP_016876881.1 | 2 | 2872 | |||
XM_017021393.1 | XP_016876882.1 | 2 | 1975 | |||
GenBank mRNA | AK000732.1 | - | 1 | 234 | ||
AK092873.1 | - | 1 | 823 | |||
AK092879.1 | - | 2 | 1632 | |||
AK225367.1 | - | 1 | 234 | |||
BC095482.1 | - | 2 | 1496 | 30334569 | ||
BC109120.1 | - | 2 | 1623 | 40035692 | ||
BC109121.1 | - | 2 | 1623 | 40035694 | ||
BX248268.1 | - | 2 | 1543 | |||
CR457249.1 | - | 1 | 228 |