Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021492.3 | NP_067467.2 | 12, 13 | 1684 | ||
GenBank mRNA | AK134504.1 | - | 12, 13 | 1433 | ||
AY528675.1 | - | 12, 13 | 1465 | |||
BC139378.1 | - | 12, 13 | 1432 | 8862400 | ||
BC139379.1 | - | 12, 13 | 1432 | 8862401 |