Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Huwe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021523.4 | NP_067498.4 | 40 | 5581 | ||
GenBank mRNA | AB093227.2 | - | 6 | 796 | ||
AY772010.3 | - | 40 | 5518 | |||
AY929611.1 | - | 38 | 5128 | |||
BC070444.1 | - | 2 | 242 | |||
DQ097265.1 | - | 40 | 5537 |