Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039126.2 | NP_001034215.1 | 4 | 1007 | ||
NM_023046.4 | NP_075533.3 | 4 | 808 | |||
GenBank mRNA | AF155352.1 | - | 4 | 759 | ||
AK006515.1 | - | 4 | 809 | |||
AK122456.1 | - | 4 | 863 | |||
AK153774.1 | - | 4 | 1029 | |||
BC034869.1 | - | 5 | 802 | |||
BC049699.1 | - | 4 | 808 | |||
KC789642.1 | - | 3 | 537 |