Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017945.3 | NP_060415.1 | 6 | 820 | ||
XM_011512948.1 | XP_011511250.1 | 6 | 670 | |||
GenBank mRNA | AK000737.1 | - | 6 | 723 | ||
AK098839.1 | - | 6 | 733 | |||
AK172825.1 | - | 1 | 26 | |||
AY358994.1 | - | 6 | 729 | |||
BC005207.1 | - | 6 | 782 | 3829330 | ||
BC010307.1 | - | 6 | 782 | 3450563 | ||
BC013046.1 | - | 6 | 724 | 3875085 |