Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025716.2 | NP_079992.1 | 2 | 791 | ||
GenBank mRNA | AK014623.1 | - | 2 | 794 | ||
AK080726.1 | - | 2 | 805 | |||
AK082745.1 | - | 2 | 805 | |||
BC014757.1 | - | 2 | 791 | 4923277 |