Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025852.3 | NP_080128.2 | 2 | 823 | ||
GenBank mRNA | AK012127.1 | - | 2 | 804 | ||
AK087016.1 | - | 2 | 808 | |||
AK144769.1 | - | 2 | 810 | |||
AK173108.1 | - | 2 | 788 | |||
BC011438.1 | - | 2 | 775 | |||
BC025511.1 | - | 2 | 794 | |||
BC039623.1 | - | 2 | 774 | |||
BC052424.1 | - | 2 | 805 | 6835200 | ||
BC054843.1 | - | 2 | 780 | |||
BC069964.1 | - | 2 | 775 |