Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxra7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026280.3 | NP_080556.1 | 4 | 1408 | ||
GenBank mRNA | AK075801.1 | - | 4 | 1409 | ||
BC147320.1 | - | 4 | 1489 | 9056373 | ||
BC147321.1 | - | 4 | 1490 | 9056376 | ||
CX734756.1 | - | 1 | 181 |