Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026331.3 | NP_080607.2 | 4 | 2250 | ||
GenBank mRNA | AF288621.1 | - | 4 | 2202 | ||
AF361699.1 | - | 3 | 3959 | |||
AK050414.1 | - | 4 | 2218 | |||
AK162615.1 | - | 2 | 2112 | |||
BC040399.1 | - | 2 | 2073 | 4037680 |