Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190179.1 | NP_001177108.1 | 8 | 869 | ||
NM_027091.4 | NP_081367.1 | 8 | 849 | |||
GenBank mRNA | AF411517.1 | - | 8 | 828 | ||
AK009187.1 | - | 8 | 797 | |||
AK089997.1 | - | 8 | 848 | |||
AK136109.1 | - | 8 | 812 | |||
AK145775.1 | - | 8 | 796 | |||
BC048814.1 | - | 8 | 804 | 2647417 |