Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027132.2 | NP_081408.2 | 6 | 1276 | ||
GenBank mRNA | AB087505.1 | - | 6 | 1174 | ||
AK043266.1 | - | 6 | 1174 | |||
BC145613.1 | - | 6 | 1136 | 9054179 | ||
BC150803.1 | - | 6 | 1136 | 9087714 |