Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027382.3 | NP_081658.1 | 4 | 510 | ||
GenBank mRNA | AK011332.1 | - | 4 | 511 | ||
AK034511.1 | - | 4 | 476 | |||
AK041965.1 | - | 4 | 509 | |||
AK131998.1 | - | 4 | 723 | |||
AY066003.1 | - | 2 | 255 | |||
BC061257.1 | - | 4 | 482 | 30251620 |