Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027551.2 | NP_081827.1 | 2 | 528 | ||
GenBank mRNA | AK086025.1 | - | 2 | 529 | ||
AK146580.1 | - | 2 | 585 | |||
AK148398.1 | - | 2 | 536 | |||
AK188689.1 | - | 1 | 66 | |||
AK199359.1 | - | 1 | 27 | |||
AK207369.1 | - | 1 | 66 | |||
BC115428.1 | - | 1 | 471 | 40094085 | ||
BC115429.1 | - | 1 | 471 | 40094088 |