Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027560.1 | NP_081836.1 | 5, 6 | 1176 | ||
GenBank mRNA | AK014582.1 | - | 5, 6 | 1176 | ||
AK046754.1 | - | 5, 6 | 886 | |||
AK135740.1 | - | 5, 6 | 992 | |||
AK137175.1 | - | 5, 6 | 993 | |||
AK144323.1 | - | 6, 7 | 910 | |||
BC036384.1 | - | 5, 6 | 974 | 1067080 | ||
BC094060.1 | - | 4, 5 | 1056 | |||
DQ861995.1 | - | 5, 6 | 840 |