Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027560.1 | NP_081836.1 | 8 | 1882 | ||
GenBank mRNA | AK014582.1 | - | 8 | 1882 | ||
AK046754.1 | - | 8 | 1592 | |||
AK137175.1 | - | 8 | 1699 | |||
AK144323.1 | - | 9 | 1616 | |||
BC034910.1 | - | 1 | 986 | |||
BC036384.1 | - | 8 | 1680 | 1067080 | ||
BC043313.1 | - | 3 | 1401 | |||
BC043469.1 | - | 3 | 1401 | |||
BC054826.1 | - | 3 | 1407 | |||
BC094060.1 | - | 7 | 1761 |