Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027861.2 | NP_082137.1 | 13 | 1274 | ||
GenBank mRNA | AK036197.1 | - | 7 | 609 | ||
AK050525.1 | - | 13 | 1245 | |||
AK146863.1 | - | 12 | 1134 | |||
AK162247.1 | - | 13 | 1244 | |||
BC030340.1 | - | 13 | 1280 | 3498026 | ||
BC120692.1 | - | 12 | 1122 | 40129615 | ||
BC125527.1 | - | 12 | 1122 | 40130042 |