Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 5 | 680 | ||
NM_001297430.1 | NP_001284359.1 | 4 | 476 | |||
NM_001297432.1 | NP_001284361.1 | 5 | 818 | |||
NM_001297433.1 | NP_001284362.1 | 5 | 754 | |||
NM_005882.4 | NP_005873.2 | 4 | 557 | |||
XM_006713849.3 | XP_006713912.1 | 5 | 640 | |||
XM_006713850.2 | XP_006713913.1 | 5 | 813 | |||
XM_011513376.1 | XP_011511678.1 | 6 | 800 | |||
XM_011513377.1 | XP_011511679.1 | 5 | 718 | |||
XM_017007636.1 | XP_016863125.1 | 6 | 800 | |||
XM_017007637.1 | XP_016863126.1 | 5 | 717 | |||
GenBank mRNA | AF084928.1 | - | 5 | 505 | ||
AK001088.1 | - | 5 | 640 | |||
AK022586.1 | - | 4 | 517 | |||
AK296174.1 | - | 4 | 433 | |||
AK298944.1 | - | 5 | 734 | |||
AK300572.1 | - | 6 | 577 | |||
AK301155.1 | - | 5 | 648 | |||
AK308901.1 | - | 4 | 501 | |||
AY189687.1 | - | 5 | 593 | |||
BC001225.1 | - | 5 | 647 | 3533990 | ||
BC006470.1 | - | 5 | 603 | |||
BT006957.1 | - | 5 | 584 |