Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027864.2 | NP_082140.2 | 15 | 2411 | ||
GenBank mRNA | AK049598.1 | - | 15 | 2357 | ||
AK078292.1 | - | 15 | 2024 | |||
BC037716.1 | - | 9 | 1390 |