Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027889.1 | NP_082165.1 | 12 | 2030 | ||
GenBank mRNA | AK004695.1 | - | 12 | 2005 | ||
AK032484.1 | - | 12 | 1995 | |||
AK037658.1 | - | 10 | 2485 | |||
AK090030.1 | - | 12 | 2011 | |||
AK153963.1 | - | 12 | 2003 | |||
AK162897.1 | - | 12 | 2005 | |||
BC029004.1 | - | 12 | 2030 | 5360111 |