Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029153.1 | NP_083429.1 | 5 | 404 | ||
GenBank mRNA | AK015706.1 | - | 4 | 365 | ||
AK082648.1 | - | 1 | 854 | |||
AK132867.1 | - | 4 | 374 | |||
AK135058.1 | - | 5 | 418 | |||
AK154671.1 | - | 5 | 418 | |||
AK162168.1 | - | 5 | 609 | |||
AK162198.1 | - | 5 | 839 | |||
AK170343.1 | - | 5 | 455 | |||
BC034283.1 | - | 5 | 404 | 4162134 |