Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052898.1 | NP_443130.1 | 3 | 3105 | ||
GenBank mRNA | AB067476.1 | - | 3 | 2639 | ||
AY534241.1 | - | 3 | 3105 |