Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052906.4 | NP_443138.2 | 3 | 3507 | ||
GenBank mRNA | AB067491.1 | - | 1 | 2726 |