Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgfac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053320.1 | NP_445772.1 | 6 | 629 | ||
XM_008770352.1 | XP_008768574.1 | 6 | 732 | |||
GenBank mRNA | BC089871.1 | - | 6 | 629 | 7324508 |