Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080442.2 | NP_536690.1 | 2 | 914 | ||
GenBank mRNA | AF201734.1 | - | 2 | 873 | ||
AF298901.1 | - | 2 | 893 | |||
AK005983.1 | - | 2 | 900 | |||
AK019840.1 | - | 2 | 915 | |||
BC048470.1 | - | 2 | 911 | 6772177 |