Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133360.2 | NP_579938.2 | 23 | 3124 | ||
GenBank mRNA | AY451393.1 | - | 23 | 3098 | ||
BC042725.1 | - | 4 | 457 | |||
CF745722.1 | - | 5 | 568 |