Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134156.2 | NP_598917.1 | 16 | 2177 | ||
GenBank mRNA | AK142591.1 | - | 7 | 893 | ||
AK154281.1 | - | 16 | 2170 | |||
BC003232.1 | - | 4 | 422 | |||
BC023206.1 | - | 3 | 379 | |||
BC054830.1 | - | 16 | 2122 | 6308412 | ||
DQ288940.1 | - | 16 | 1897 |