Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134156.2 | NP_598917.1 | 20 | 2747 | ||
GenBank mRNA | AK014073.1 | - | 1 | 55 | ||
AK142591.1 | - | 11 | 1464 | |||
AK154281.1 | - | 20 | 2740 | |||
BC003232.1 | - | 8 | 992 | |||
BC038919.1 | - | 3 | 602 | |||
BC054830.1 | - | 20 | 2692 | 6308412 | ||
DQ288940.1 | - | 20 | 2452 |