Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146032.1 | NP_001139504.1 | 25 | 2972 | ||
NM_138782.2 | NP_620137.2 | 26 | 3071 | |||
XM_017009016.1 | XP_016864505.1 | 25 | 2950 | |||
GenBank mRNA | AK293796.1 | - | 10 | 1714 | ||
AL831971.1 | - | 26 | 2980 | |||
AL832928.1 | - | 8 | 1412 | |||
AL833313.1 | - | 23 | 2670 | |||
BC137070.1 | - | 26 | 3071 | 9021067 |