Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146032.1 | NP_001139504.1 | 19 | 1645 | ||
NM_138782.2 | NP_620137.2 | 20 | 1744 | |||
XM_017009016.1 | XP_016864505.1 | 19 | 1623 | |||
XM_017009017.1 | XP_016864506.1 | 20 | 1738 | |||
XM_017009018.1 | XP_016864507.1 | 20 | 1738 | |||
XM_017009020.1 | XP_016864509.1 | 19 | 1634 | |||
XM_017009021.1 | XP_016864510.1 | 19 | 1634 | |||
GenBank mRNA | AK291782.1 | - | 20 | 1674 | ||
AK293796.1 | - | 4 | 387 | |||
AK295141.1 | - | 19 | 1555 | |||
AL831971.1 | - | 20 | 1653 | |||
AL832928.1 | - | 2 | 85 | |||
AL833313.1 | - | 17 | 1343 | |||
BC137070.1 | - | 20 | 1744 | 9021067 |