Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139230.1 | NP_631976.1 | 4 | 398 | ||
NM_139231.1 | NP_631977.1 | 3 | 206 | |||
XM_006233479.2 | XP_006233541.1 | 5 | 524 | |||
XM_006233480.2 | XP_006233542.1 | 5 | 773 | |||
XM_006233483.2 | XP_006233545.1 | 5 | 524 | |||
XM_006233484.2 | XP_006233546.1 | 4 | 331 | |||
XM_006233485.2 | XP_006233547.1 | 4 | 331 | |||
XM_008761548.1 | XP_008759770.1 | 4 | 473 | |||
GenBank mRNA | AF056034.1 | - | 4 | 398 | ||
AF056035.1 | - | 3 | 206 |