Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145439.2 | NP_663414.3 | 6 | 679 | ||
NM_181321.3 | NP_851838.1 | 6 | 617 | |||
GenBank mRNA | AK052814.1 | - | 6 | 619 | ||
AK077671.1 | - | 5 | 521 | |||
AK090179.1 | - | 6 | 636 | |||
AY236497.1 | - | 6 | 578 | |||
AY263158.1 | - | 6 | 604 | |||
BC004840.1 | - | 6 | 604 | 2648849 | ||
BC006956.1 | - | 4 | 389 | |||
BC058195.1 | - | 6 | 604 | 5356348 |