Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145968.2 | NP_666080.1 | 10 | 2903 | ||
GenBank mRNA | BC030886.1 | - | 10 | 2883 | 4483459 | |
BC039780.1 | - | 10 | 2894 | 4483459 |
Gene Symbol: Tagap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147155.2 | NP_671511.2 | 4 | 2139 | ||
GenBank mRNA | AK012033.1 | - | 1 | 1375 |