Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013666.2 | NP_038694.2 | 5, 6 | 820 | ||
NM_153124.2 | NP_694764.2 | 4, 5 | 712 | |||
GenBank mRNA | AK078670.1 | - | 4, 5 | 495 | ||
AK162549.1 | - | 4, 5 | 492 | |||
BC034855.1 | - | 4, 5 | 706 | 5146703 | ||
X98014.1 | - | 5, 6 | 683 |