Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027321.3 | NP_081597.1 | 5 | 633 | ||
NM_175413.3 | NP_780622.1 | 5 | 633 | |||
GenBank mRNA | AK008119.1 | - | 5 | 601 | ||
AK035072.1 | - | 5 | 635 | |||
AK039277.1 | - | 5 | 630 | |||
AK046671.1 | - | 5 | 529 | |||
AK052159.1 | - | 5 | 585 | |||
AK079125.1 | - | 5 | 611 | |||
BC119529.2 | - | 5 | 582 | 40056120 | ||
BC119530.1 | - | 5 | 583 | 40056121 |