Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029512.2 | NP_083788.2 | 5 | 2353 | ||
NM_181734.3 | NP_859423.2 | 5 | 2350 | |||
GenBank mRNA | AK081680.1 | - | 5 | 2313 | ||
AK156641.1 | - | 5 | 2316 | |||
BC052389.1 | - | 5 | 2308 | 5718874 | ||
BC060726.1 | - | 4 | 2219 |