Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029512.2 | NP_083788.2 | 5 | 2527 | ||
NM_181734.3 | NP_859423.2 | 5 | 2524 | |||
GenBank mRNA | AK081680.1 | - | 5 | 2487 | ||
AK156641.1 | - | 5 | 2490 | |||
BC052389.1 | - | 5 | 2482 | 5718874 | ||
BC060726.1 | - | 4 | 2393 |