Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318781.1 | NP_001305710.1 | 36, 37 | 5569 | ||
NM_182920.1 | NP_891550.1 | 37, 38 | 5653 | |||
GenBank mRNA | AF488803.1 | - | 37, 38 | 5653 | ||
AK300357.1 | - | 16, 17 | 2325 | |||
AK303343.1 | - | 18, 19 | 2788 | |||
BC130578.1 | - | 37, 38 | 5643 | 40146609 | ||
BC171764.1 | - | 36, 37 | 5559 | 9054417 |