Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207176.3 | NP_997059.1 | 7 | 1421 | ||
GenBank mRNA | AK132249.1 | - | 7 | 1379 | ||
AK145609.1 | - | 7 | 1370 | |||
AK167101.1 | - | 7 | 1370 | |||
AK167274.1 | - | 7 | 1378 | |||
BC003808.1 | - | 9 | 1661 | 3589585 | ||
BC010465.1 | - | 7 | 1377 | 3585670 | ||
BC051103.1 | - | 7 | 1353 | |||
X78989.1 | - | 6 | 1250 |