Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207176.3 | NP_997059.1 | 5 | 901 | ||
GenBank mRNA | AK132249.1 | - | 5 | 859 | ||
AK145609.1 | - | 5 | 850 | |||
AK167101.1 | - | 5 | 850 | |||
AK167274.1 | - | 5 | 858 | |||
BC003808.1 | - | 7 | 1141 | 3589585 | ||
BC010465.1 | - | 5 | 857 | 3585670 | ||
BC051103.1 | - | 5 | 835 | |||
CT010319.1 | - | 4 | 736 | |||
X78989.1 | - | 4 | 730 |