Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153696.2 | NP_710163.1 | 1 | 13 | ||
GenBank mRNA | AF261715.1 | - | 1 | 13 |