Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318781.1 | NP_001305710.1 | 39 | 7081 | ||
NM_182920.1 | NP_891550.1 | 40 | 7165 | |||
GenBank mRNA | AK124945.1 | - | 2 | 1666 | ||
BC026271.2 | - | 3 | 1523 |