Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009618.1 | NP_001009618.1 | 5 | 497 | ||
XM_006232043.2 | XP_006232105.1 | 5 | 464 | |||
GenBank mRNA | BC088346.1 | - | 5 | 497 | 7368325 | |
FQ211681.1 | - | 5 | 454 | |||
FQ211748.1 | - | 5 | 454 | |||
FQ228567.1 | - | 2 | 183 | |||
FQ230867.1 | - | 5 | 427 | |||
FQ232387.1 | - | 5 | 426 | |||
K02816.1 | - | 4 | 352 |