Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009685.1 | NP_001009685.1 | 7 | 879 | ||
GenBank mRNA | BC088410.1 | - | 7 | 879 | 7189810 |