Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009696.1 | NP_001009696.1 | 7 | 859 | ||
GenBank mRNA | BC087627.1 | - | 7 | 859 | 7318159 |